Product Details
- SNP ID
-
rs201590585
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149071519 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGAACGGTCTCTCCCCACCGTGC[A/G]CGTGCTGGTGGACCTGCAGCAGGCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF786
PubMed Links
Gene Details
- Gene
- ZNF786
- Gene Name
- zinc finger protein 786
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152411.3 |
1390 |
Missense Mutation |
GCG,GTG |
A418V |
NP_689624.2 |
View Full Product Details