Product Details

SNP ID
rs202094370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100402517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTTCCAGCTGGCCTGCTTACC[C/T]CTTGGCTTGGTTTTTTGGGTCTGTA
Phenotype
MIM: 605341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 2284 Intron NP_001244937.1
NM_017984.4 2284 Silent Mutation AGA,AGG R490R NP_060454.3
XM_005250479.2 2284 Intron XP_005250536.1
XM_005250480.2 2284 Intron XP_005250537.1
XM_006716035.3 2284 Silent Mutation AGA,AGG R492R XP_006716098.1
XM_006716036.2 2284 Silent Mutation AGA,AGG R492R XP_006716099.1
XM_006716037.3 2284 Silent Mutation AGA,AGG R491R XP_006716100.1
XM_006716038.3 2284 Silent Mutation AGA,AGG R491R XP_006716101.1
XM_006716040.3 2284 Silent Mutation AGA,AGG R465R XP_006716103.1
XM_017012379.1 2284 Intron XP_016867868.1
XM_017012380.1 2284 Intron XP_016867869.1

View Full Product Details