Product Details

SNP ID
rs202150910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:54542782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTTGTTTTCTTTTCCGTTTTA[G/T]ATGTACAACAAGGGCTTTCTTCTCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VSTM2A PubMed Links

Gene Details

Gene
VSTM2A
Gene Name
V-set and transmembrane domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301009.1 458 Missense Mutation GAT,TAT D18Y NP_001287938.1
NM_001317843.1 458 Missense Mutation GAT,TAT D18Y NP_001304772.1
NM_182546.3 458 Missense Mutation GAT,TAT D18Y NP_872352.2
XM_006715665.3 458 Missense Mutation GAT,TAT D18Y XP_006715728.1
XM_006715666.3 458 Missense Mutation GAT,TAT D18Y XP_006715729.1
XM_017011841.1 458 Missense Mutation GAT,TAT D18Y XP_016867330.1

View Full Product Details