Product Details

SNP ID
rs201480496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19116755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCAGGCCCCCTCCATCCTCCA[C/G]ACCGAGAAGGCGTAGCTGAGCCGCT
Phenotype
MIM: 601622
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TWIST1 PubMed Links

Gene Details

Gene
TWIST1
Gene Name
twist family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000474.3 665 Silent Mutation GTC,GTG V189V NP_000465.1
XM_011515496.1 665 Silent Mutation GTC,GTG V189V XP_011513798.1

View Full Product Details