Product Details

SNP ID
rs202237202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100374291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTTTCTGAACTGGACAGAGGG[C/T]CAGAAGAGCGGCTTCCTCAGGATCT
Phenotype
MIM: 605341 MIM: 605342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013439.2 524 Silent Mutation GGC,GGT G104G NP_038467.2
NM_178272.1 524 Silent Mutation GGC,GGT G104G NP_840056.1
NM_178273.1 524 Silent Mutation GGC,GGT G104G NP_840057.1
Gene
PILRB
Gene Name
paired immunoglobin-like type 2 receptor beta
There are no transcripts associated with this gene.

Gene
STAG3L5P-PVRIG2P-PILRB
Gene Name
STAG3L5P-PVRIG2P-PILRB readthrough
There are no transcripts associated with this gene.

View Full Product Details