Product Details

SNP ID
rs201593102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74096750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCTCTGAGCAAATCACCAAGG[C/G]ACTGGTTATGGTGAGCGCCCCCTGC
Phenotype
MIM: 601329
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LIMK1 PubMed Links

Gene Details

Gene
LIMK1
Gene Name
LIM domain kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204426.1 273 Missense Mutation GCA,GGA A60G NP_001191355.1
NM_002314.3 273 Missense Mutation GCA,GGA A94G NP_002305.1

View Full Product Details