Product Details

SNP ID
rs202099108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108884451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGAAACAGCATTGCAAGGCAAC[C/T]GAGGCTCCAAAGTCTCTGATGAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf66 PubMed Links

Gene Details

Gene
C7orf66
Gene Name
chromosome 7 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024607.2 137 Missense Mutation AGT,GGT S28G NP_001019778.1

View Full Product Details