Product Details

SNP ID
rs201501752
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94663817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGACCTCCCAGAGAAGTTCGA[C/T]GGCAACCCAGACATGCTGGCTCCTT
Phenotype
MIM: 609810 MIM: 604149
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PEG10 PubMed Links

Gene Details

Gene
PEG10
Gene Name
paternally expressed 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040152.1 740 Silent Mutation GAC,GAT D87D NP_001035242.1
NM_001172437.2 740 Silent Mutation GAC,GAT D163D NP_001165908.1
NM_001172438.2 740 Silent Mutation GAC,GAT D163D NP_001165909.1
NM_001184961.1 740 Silent Mutation GAC,GAT D121D NP_001171890.1
NM_001184962.1 740 Silent Mutation GAC,GAT D121D NP_001171891.1
NM_015068.3 740 Silent Mutation GAC,GAT D87D NP_055883.2
Gene
SGCE
Gene Name
sarcoglycan epsilon
There are no transcripts associated with this gene.

View Full Product Details