Product Details

SNP ID
rs199602591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143026784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTACACACAGCCACATAACGGT[C/T]CACAGCCATCACTCCAAGTAATGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9A2 PubMed Links

Gene Details

Gene
OR9A2
Gene Name
olfactory receptor family 9 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001658.1 349 Missense Mutation AAC,GAC N117D NP_001001658.1

View Full Product Details