Product Details

SNP ID
rs199622831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100401046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACAGCGCCACGGGGAAGTCCT[C/T]GCCATCACTGTTGCTGTGCTGCAGC
Phenotype
MIM: 605341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013439.2 2729 Intron NP_038467.2
NM_178272.1 2729 Intron NP_840056.1
NM_178273.1 2729 Intron NP_840057.1
Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 2729 UTR 3 NP_001244937.1
NM_017984.4 2729 Missense Mutation AAG,GAG K639E NP_060454.3
XM_005250479.2 2729 UTR 3 XP_005250536.1
XM_005250480.2 2729 UTR 3 XP_005250537.1
XM_006716035.3 2729 Missense Mutation AAG,GAG K641E XP_006716098.1
XM_006716036.2 2729 Missense Mutation AAG,GAG K641E XP_006716099.1
XM_006716037.3 2729 Missense Mutation AAG,GAG K640E XP_006716100.1
XM_006716038.3 2729 Missense Mutation AAG,GAG K640E XP_006716101.1
XM_006716040.3 2729 Missense Mutation AAG,GAG K614E XP_006716103.1
XM_017012379.1 2729 UTR 3 XP_016867868.1
XM_017012380.1 2729 UTR 3 XP_016867869.1

View Full Product Details