Product Details

SNP ID
rs199570229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100155144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACAGGGGGCGTGATGGCTCTG[C/T]GCTCCATCACACTGCCCGACCTGGG
Phenotype
MIM: 608235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf43 PubMed Links

Gene Details

Gene
C7orf43
Gene Name
chromosome 7 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303470.1 1332 Missense Mutation CAC,CGC H268R NP_001290399.1
NM_018275.4 1332 Missense Mutation CAC,CGC H537R NP_060745.3
Gene
GAL3ST4
Gene Name
galactose-3-O-sulfotransferase 4
There are no transcripts associated with this gene.

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008395.3 1332 Intron NP_001008396.1
NM_001318236.1 1332 Intron NP_001305165.1
NM_001318237.1 1332 Intron NP_001305166.1
XM_017012198.1 1332 Intron XP_016867687.1
XM_017012199.1 1332 Intron XP_016867688.1
Gene
MIR4658
Gene Name
microRNA 4658
There are no transcripts associated with this gene.

View Full Product Details