Product Details

SNP ID
rs199703984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73865625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAGCTCCTGGTACAGCTGAGA[C/T]GTCTGTATTGGTGGGTGGAGACTAT
Phenotype
MIM: 612547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WBSCR28 PubMed Links

Gene Details

Gene
WBSCR28
Gene Name
Williams-Beuren syndrome chromosome region 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182504.3 586 Missense Mutation CGT,TGT R184C NP_872310.2
XM_011515785.2 586 Missense Mutation CGT,TGT R87C XP_011514087.1
XM_017011741.1 586 Missense Mutation CGT,TGT R87C XP_016867230.1

View Full Product Details