Product Details

SNP ID
rs199687267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117368657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGTAATAACATTCTGTACAGC[C/T]GTTATTAAATGGCCACACTGTTTAT
Phenotype
MIM: 605797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASZ1 PubMed Links

Gene Details

Gene
ASZ1
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301821.1 1067 Silent Mutation ACA,ACG T372T NP_001288750.1
NM_001301822.1 1067 Silent Mutation ACA,ACG T164T NP_001288751.1
NM_130768.2 1067 Silent Mutation ACA,ACG T372T NP_570124.1

View Full Product Details