Product Details
- SNP ID
-
rs199706346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:111728425 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGACGGGGATGGAGAGGCTGTGA[A/G]GTAGCGGGACGGGGCGCCGCAGAGT
- Phenotype
-
MIM: 607679
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DOCK4
PubMed Links
Gene Details
- Gene
- DOCK4
- Gene Name
- dedicator of cytokinesis 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014705.3 |
6551 |
Missense Mutation |
CCT,CTT |
P1917L |
NP_055520.3 |
XM_006716188.2 |
6551 |
Missense Mutation |
CCT,CTT |
P1926L |
XP_006716251.1 |
XM_006716189.2 |
6551 |
Missense Mutation |
CCT,CTT |
P1888L |
XP_006716252.1 |
XM_017012819.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1957L |
XP_016868308.1 |
XM_017012820.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1948L |
XP_016868309.1 |
XM_017012821.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1939L |
XP_016868310.1 |
XM_017012822.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1937L |
XP_016868311.1 |
XM_017012823.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1919L |
XP_016868312.1 |
XM_017012824.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1910L |
XP_016868313.1 |
XM_017012825.1 |
6551 |
Missense Mutation |
CCT,CTT |
P1901L |
XP_016868314.1 |
View Full Product Details