Product Details

SNP ID
rs199822756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150340140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCACTCGGGTTTCTTCCAGTCC[C/T]TCTTCCGGCAGCTTGTCTGCTGCAG
Phenotype
MIM: 601973 MIM: 615252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC61 PubMed Links

Gene Details

Gene
LRRC61
Gene Name
leucine rich repeat containing 61
There are no transcripts associated with this gene.

Gene
RARRES2
Gene Name
retinoic acid receptor responder 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002889.3 356 Missense Mutation NP_002880.1
XM_017012491.1 356 Intron XP_016867980.1
Gene
ZBED6CL
Gene Name
ZBED6 C-terminal like
There are no transcripts associated with this gene.

View Full Product Details