Product Details

SNP ID
rs199829808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143222154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACCTTCACTTTGGTGGTCTCC[A/G]GAATAGAGTGCATCACTGGCATCCT
Phenotype
MIM: 613964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R40 PubMed Links

Gene Details

Gene
TAS2R40
Gene Name
taste 2 receptor member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176882.1 76 Missense Mutation AGA,GGA R26G NP_795363.1

View Full Product Details