Product Details

SNP ID
rs199847662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100467139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCACCTCCACCTCCTCCCGGAC[C/T]GCAAACATGAGGTGGGACTTCACCA
Phenotype
MIM: 611914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf61 PubMed Links

Gene Details

Gene
C7orf61
Gene Name
chromosome 7 open reading frame 61
There are no transcripts associated with this gene.

Gene
TSC22D4
Gene Name
TSC22 domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303043.1 1763 Silent Mutation GCA,GCG A336A NP_001289972.1
NM_030935.4 1763 Silent Mutation GCA,GCG A336A NP_112197.1

View Full Product Details