Product Details
- SNP ID
-
rs199793084
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141664177 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACGGCCCCAGGACTCACAGCAC[C/G]AAGAGGTCCTCTTCACAAGGGTTGT
- Phenotype
-
MIM: 610345
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AGK
PubMed Links
Gene Details
- Gene
- AGK
- Gene Name
- acylglycerol kinase
There are no transcripts associated with this gene.
- Gene
- KIAA1147
- Gene Name
- KIAA1147
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080392.1 |
1167 |
Silent Mutation |
TTC,TTG |
F389L |
NP_001073861.1 |
View Full Product Details