Product Details

SNP ID
rs2229779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90060698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTCATCTATGTATCCATTGCC[G/A]TCCTGGGGGAGGAGAAATAAAATAG
Phenotype
MIM: 114050 MIM: 222745
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CALB1 PubMed Links

Gene Details

Gene
CALB1
Gene Name
calbindin 1
There are no transcripts associated with this gene.

Gene
DECR1
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details