Product Details
- SNP ID
-
hCV191897243
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:81585647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCCCCGGTGGGGCAGTACCCCAG[G/A]GAGAAGATCTACAAGGAGCAAGACA
- Phenotype
-
MIM: 600189
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLE1
PubMed Links
Gene Details
- Gene
- TLE1
- Gene Name
- transducin like enhancer of split 1
There are no transcripts associated with this gene.
View Full Product Details