Product Details

SNP ID
rs201563373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144393507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTCGCTGCGCTCCATGGTGCTC[A/G]GGTACAGGTAGCGGTTGAGCAGCTC
Phenotype
MIM: 606027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCK5 PubMed Links

Gene Details

Gene
ADCK5
Gene Name
aarF domain containing kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174922.4 4254 Intron NP_777582.4
XM_006716527.2 4254 Intron XP_006716590.1
XM_011516907.2 4254 Intron XP_011515209.1
XM_011516909.1 4254 Intron XP_011515211.1
XM_011516910.1 4254 Intron XP_011515212.1
XM_017013174.1 4254 Intron XP_016868663.1
Gene
CPSF1
Gene Name
cleavage and polyadenylation specific factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013291.2 4254 Missense Mutation CCG,CTG P1410L NP_037423.2
XM_006716548.2 4254 Missense Mutation CCG,CTG P1407L XP_006716611.1
XM_006716549.2 4254 Missense Mutation CCG,CTG P1332L XP_006716612.1
XM_006716550.3 4254 Missense Mutation CCG,CTG P752L XP_006716613.1
XM_011516997.1 4254 Missense Mutation CCG,CTG P1332L XP_011515299.1
XM_011516998.1 4254 Missense Mutation CCG,CTG P1332L XP_011515300.1
XM_011516999.1 4254 Missense Mutation CCG,CTG P1332L XP_011515301.1
Gene
MIR1234
Gene Name
microRNA 1234
There are no transcripts associated with this gene.

Gene
MIR6849
Gene Name
microRNA 6849
There are no transcripts associated with this gene.

Gene
MIR939
Gene Name
microRNA 939
There are no transcripts associated with this gene.

View Full Product Details