Product Details

SNP ID
rs201817819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109243185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGTTGTAAAGTACAGTGGACA[G/T]GGGAGCAGGCTGACGATAGATGAAT
Phenotype
MIM: 606109
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NUDCD1 PubMed Links

Gene Details

Gene
NUDCD1
Gene Name
NudC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128211.1 1587 Missense Mutation NP_001121683.1
NM_032869.3 1587 Missense Mutation NP_116258.2
XM_017013910.1 1587 Missense Mutation XP_016869399.1
XM_017013911.1 1587 Missense Mutation XP_016869400.1

View Full Product Details