Product Details
- SNP ID
-
rs201250823
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143693890 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTGCTCACGCAGCGTTGTGGGC[A/G]GCGGCCGGGCCGCAGAGAGCGCCGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BREA2
PubMed Links
Gene Details
- Gene
- BREA2
- Gene Name
- breast cancer estrogen-induced apoptosis 2
There are no transcripts associated with this gene.
- Gene
- ZNF707
- Gene Name
- zinc finger protein 707
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001100598.1 |
717 |
Missense Mutation |
CAG,CGG |
Q159R |
NP_001094068.1 |
NM_001100599.1 |
717 |
Missense Mutation |
CAG,CGG |
Q159R |
NP_001094069.1 |
NM_001288805.1 |
717 |
Missense Mutation |
CAG,CGG |
Q159R |
NP_001275734.1 |
NM_001288806.1 |
717 |
Missense Mutation |
CAG,CGG |
Q159R |
NP_001275735.1 |
NM_001288807.1 |
717 |
Missense Mutation |
CAG,CGG |
Q86R |
NP_001275736.1 |
NM_001288808.1 |
717 |
Missense Mutation |
CAG,CGG |
Q123R |
NP_001275737.1 |
NM_001288809.1 |
717 |
Missense Mutation |
CAG,CGG |
Q123R |
NP_001275738.1 |
NM_173831.3 |
717 |
Missense Mutation |
CAG,CGG |
Q159R |
NP_776192.2 |
XM_005250883.3 |
717 |
Missense Mutation |
CAG,CGG |
Q86R |
XP_005250940.1 |
XM_006716542.3 |
717 |
Missense Mutation |
CAG,CGG |
Q86R |
XP_006716605.1 |
XM_006716543.3 |
717 |
Missense Mutation |
CAG,CGG |
Q86R |
XP_006716606.1 |
XM_011516977.2 |
717 |
Missense Mutation |
CAG,CGG |
Q159R |
XP_011515279.1 |
XM_011516979.2 |
717 |
Missense Mutation |
CAG,CGG |
Q86R |
XP_011515281.1 |
XM_017013335.1 |
717 |
Missense Mutation |
CAG,CGG |
Q86R |
XP_016868824.1 |
View Full Product Details