Product Details

SNP ID
rs201693505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:108203109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGACTGCATTGTTACCCATAAC[C/T]ACATGACCCTAAAAGGAAACACAGG
Phenotype
MIM: 602210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3E PubMed Links

Gene Details

Gene
EIF3E
Gene Name
eukaryotic translation initiation factor 3 subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001568.2 1273 Intron NP_001559.1
XM_017013398.1 1273 Silent Mutation GTA,GTG V419V XP_016868887.1

View Full Product Details