Product Details

SNP ID
rs201821196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10898151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGCCCTTCTGGGAGGTAAGGA[C/T]GCCCCAACGGGGTAGGGGGCCCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XKR6 PubMed Links

Gene Details

Gene
XKR6
Gene Name
XK related 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173683.3 1754 Missense Mutation CAT,CGT H576R NP_775954.2
XM_011543821.2 1754 Intron XP_011542123.1

View Full Product Details