Product Details
- SNP ID
-
rs201609532
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:132024530 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCAAGGGACCCAGTGACTTCCGC[A/G]GAAACCTCTTGGCTCTGCCGAGGTC
- Phenotype
-
MIM: 601658
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OC90
PubMed Links
Gene Details
- Gene
- OC90
- Gene Name
- otoconin 90
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080399.2 |
1472 |
Missense Mutation |
CCG,CTG |
P462L |
NP_001073868.2 |
View Full Product Details