Product Details

SNP ID
rs202056172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142727451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGTGCGCCGTGCTGCGCGCGCG[C/T]CTGCTGCAGCCGCGCGTCCGTGACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
THEM6
Gene Name
thioesterase superfamily member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016647.2 249 Silent Mutation CGC,CGT R35R NP_057731.1
XM_005250955.2 249 Silent Mutation CGC,CGT R35R XP_005251012.1

View Full Product Details