Product Details

SNP ID
rs202131264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:54135288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTCTTGGATCTGCGGGTTATGG[C/T]CGGTCCCTTGCAGGGCGGTGGGGCC
Phenotype
MIM: 611828
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL15 PubMed Links

Gene Details

Gene
MRPL15
Gene Name
mitochondrial ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014175.3 79 Missense Mutation GCC,GTC A2V NP_054894.1
XM_017013362.1 79 Missense Mutation GCC,GTC A2V XP_016868851.1

View Full Product Details