Product Details
- SNP ID
-
rs200979468
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:81694320 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGCAAAGAGATAGCCCCCACCC[C/G]CTCCTGGGCACGTGCAGGTCATTAC
- Phenotype
-
MIM: 602064
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IMPA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75192971] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IMPA1
- Gene Name
- inositol monophosphatase 1
There are no transcripts associated with this gene.
- Gene
- SLC10A5
- Gene Name
- solute carrier family 10 member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010893.2 |
653 |
Missense Mutation |
GCG,GGG |
A218G |
NP_001010893.1 |
- Gene
- ZFAND1
- Gene Name
- zinc finger AN1-type containing 1
There are no transcripts associated with this gene.
View Full Product Details