Product Details

SNP ID
rs201149309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23433081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACCAGGAAGCAGCCAGAGAACA[C/G]GTAGTGGTTGTAGACAAAGGAAACG
Phenotype
MIM: 607577
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ENTPD4 PubMed Links

Gene Details

Gene
ENTPD4
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128930.2 1944 Missense Mutation CTG,GTG L558V NP_001122402.1
NM_004901.4 1944 Missense Mutation CTG,GTG L566V NP_004892.1

View Full Product Details