Product Details

SNP ID
rs200936938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30680922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTACCTTTTCTTCCTTGTTAGC[A/G]CAGACCATTTTCATCACACATTTTG
Phenotype
MIM: 138300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSR PubMed Links

Gene Details

Gene
GSR
Gene Name
glutathione-disulfide reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000637.3 1289 Silent Mutation TGC,TGT C467C NP_000628.2
NM_001195102.1 1289 Silent Mutation TGC,TGT C438C NP_001182031.1
NM_001195103.1 1289 Silent Mutation TGC,TGT C414C NP_001182032.1
NM_001195104.1 1289 Silent Mutation TGC,TGT C385C NP_001182033.1

View Full Product Details