Product Details

SNP ID
rs201763716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:106761408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGAAAGGATTGAGCTTCTGGG[A/G]TTGTATGTGTTCATCAGCCCACTGC
Phenotype
MIM: 609747 MIM: 605609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABRA PubMed Links

Gene Details

Gene
ABRA
Gene Name
actin binding Rho activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139166.4 829 Missense Mutation CCC,TCC P259S NP_631905.1
Gene
OXR1
Gene Name
oxidation resistance 1
There are no transcripts associated with this gene.

View Full Product Details