Product Details

SNP ID
rs202146006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27287594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACACTGATGTGCAAGGGGCAGA[C/T]GCGCAAAGGCTCTGGAGGCCCGGCG
Phenotype
MIM: 617007
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TRIM35 PubMed Links

Gene Details

Gene
TRIM35
Gene Name
tripartite motif containing 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304495.1 1550 UTR 3 NP_001291424.1
NM_171982.4 1550 Missense Mutation ATC,GTC I480V NP_741983.2
XM_005273452.3 1550 UTR 3 XP_005273509.1
XM_011544453.1 1550 Missense Mutation ATC,GTC I330V XP_011542755.1

View Full Product Details