Product Details

SNP ID
rs201152934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:19819621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGGTGTGGAGAGAAATCAGCAT[C/T]ATTACATCAGCATTAAGGAACGATT
Phenotype
MIM: 611353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INTS10 PubMed Links

Gene Details

Gene
INTS10
Gene Name
integrator complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018142.2 200 Silent Mutation ATC,ATT I82I NP_060612.2
XM_005273555.3 200 Silent Mutation ATC,ATT I82I XP_005273612.1
XM_005273556.3 200 Silent Mutation ATC,ATT I82I XP_005273613.1
XM_005273557.3 200 Silent Mutation ATC,ATT I82I XP_005273614.1
XM_006716355.2 200 UTR 5 XP_006716418.1
XM_017013606.1 200 Silent Mutation ATC,ATT I82I XP_016869095.1
XM_017013607.1 200 Silent Mutation ATC,ATT I82I XP_016869096.1
XM_017013608.1 200 UTR 5 XP_016869097.1
XM_017013609.1 200 UTR 5 XP_016869098.1

View Full Product Details