Product Details

SNP ID
rs201301380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10765516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCTCTCCTCTTTTTGGGCTTC[A/G]GGGTGAAGTCCCGGCCCTCAGGCGG
Phenotype
MIM: 606505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102723313 PubMed Links

Gene Details

Gene
LOC102723313
Gene Name
uncharacterized LOC102723313
There are no transcripts associated with this gene.

Gene
PINX1
Gene Name
PIN2/TERF1 interacting, telomerase inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details