Product Details
- SNP ID
-
rs201222374
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:142703102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTTCCTATTAGAAATATTTCCA[C/T]GTTTTTTCATGGTTGCGAAGCAGTG
- Phenotype
-
MIM: 615093
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100288181
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2244163] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100288181
- Gene Name
- uncharacterized LOC100288181
There are no transcripts associated with this gene.
- Gene
- LY6K
- Gene Name
- lymphocyte antigen 6 complex, locus K
View Full Product Details