Product Details
- SNP ID
-
rs201855218
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143916574 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCTGTGCGGTGCGGGCGTCCACC[A/G]TGCCGCGCTGCAGGGCCTCGTCCAG
- Phenotype
-
MIM: 601282
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLEC
PubMed Links
Gene Details
- Gene
- PLEC
- Gene Name
- plectin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000445.4 |
13193 |
Missense Mutation |
ACG,ATG |
T4443M |
NP_000436.2 |
NM_201378.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4402M |
NP_958780.1 |
NM_201379.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4394M |
NP_958781.1 |
NM_201380.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4553M |
NP_958782.1 |
NM_201381.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4384M |
NP_958783.1 |
NM_201382.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4416M |
NP_958784.1 |
NM_201383.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4420M |
NP_958785.1 |
NM_201384.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4416M |
NP_958786.1 |
XM_005250976.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4558M |
XP_005251033.1 |
XM_005250978.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4425M |
XP_005251035.1 |
XM_005250979.4 |
13193 |
Missense Mutation |
ACG,ATG |
T4421M |
XP_005251036.1 |
XM_005250980.4 |
13193 |
Missense Mutation |
ACG,ATG |
T4421M |
XP_005251037.1 |
XM_005250981.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4407M |
XP_005251038.1 |
XM_005250982.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4399M |
XP_005251039.1 |
XM_005250983.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4393M |
XP_005251040.1 |
XM_005250984.4 |
13193 |
Missense Mutation |
ACG,ATG |
T4389M |
XP_005251041.1 |
XM_006716588.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4448M |
XP_006716651.1 |
XM_006716589.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4398M |
XP_006716652.1 |
XM_006716590.3 |
13193 |
Missense Mutation |
ACG,ATG |
T4398M |
XP_006716653.1 |
XM_011517130.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4421M |
XP_011515432.1 |
XM_011517131.2 |
13193 |
Missense Mutation |
ACG,ATG |
T4393M |
XP_011515433.1 |
XM_011517132.2 |
13193 |
Missense Mutation |
ACG,ATG |
T3298M |
XP_011515434.1 |
View Full Product Details