Product Details

SNP ID
rs200945105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10525640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCTGGTCACGGGAACTCAGCTC[A/G]GTCCACGGACTCCTCTCCCAGAGGC
Phenotype
MIM: 615144
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS55 PubMed Links

Gene Details

Gene
PRSS55
Gene Name
protease, serine 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197020.1 310 Missense Mutation AGT,GGT S19G NP_001183949.1
NM_198464.3 310 Missense Mutation AGT,GGT S19G NP_940866.2
XM_005272372.3 310 Missense Mutation AGT,GGT S84G XP_005272429.2
XM_011543814.2 310 Missense Mutation AGT,GGT S84G XP_011542116.2
XM_017013182.1 310 Missense Mutation AGT,GGT S84G XP_016868671.1

View Full Product Details