Product Details

SNP ID
rs201709007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:97775789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTCCGCCCCGCCCCCTCCCCG[C/T]CCCCGCCGCTGCAGCGGTCGCCTTC
Phenotype
MIM: 613296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAPTM4B PubMed Links

Gene Details

Gene
LAPTM4B
Gene Name
lysosomal protein transmembrane 4 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018407.4 209 Missense Mutation GCC,GTC A18V NP_060877.3

View Full Product Details