Product Details

SNP ID
rs201814058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144096223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGTGGGCGTGTGCTACACGGA[G/T]GATGAAGCTAAGGAGCTGGCTGCGG
Phenotype
MIM: 123980 MIM: 610210 MIM: 611885
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYC1 PubMed Links

Gene Details

Gene
CYC1
Gene Name
cytochrome c1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001916.4 491 Silent Mutation GAG,GAT E142D NP_001907.2
XM_017013102.1 491 Silent Mutation GAG,GAT E83D XP_016868591.1
Gene
MAF1
Gene Name
MAF1 homolog, negative regulator of RNA polymerase III
There are no transcripts associated with this gene.

Gene
SHARPIN
Gene Name
SHANK associated RH domain interactor
There are no transcripts associated with this gene.

View Full Product Details