Product Details
- SNP ID
-
rs202180381
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:37965318 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCTGGGCTGCTCCGGGCCGCAGG[A/C]ACGCCCGAGGGGAAGAGGGCCGGGC
- Phenotype
-
MIM: 109691
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADRB3
PubMed Links
Gene Details
- Gene
- ADRB3
- Gene Name
- adrenoceptor beta 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000025.2 |
1349 |
Silent Mutation |
GTG,GTT |
V384V |
NP_000016.1 |
View Full Product Details