Product Details
- SNP ID
-
rs200926173
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:93728244 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTAAGAAAGGATCAACAAGCAGAA[C/G]ATGATGAGGATGACGAGTTAGATGT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM92A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147392141] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM92A1
- Gene Name
- family with sequence similarity 92 member A1
- Gene
- RBM12B
- Gene Name
- RNA binding motif protein 12B
View Full Product Details