Product Details

SNP ID
rs201350341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38991431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACCAAATCCCAAGTCCTCCAAG[C/T]GTCAACAGCTTCCCTACTGCAGTGC
Phenotype
MIM: 602713 MIM: 610700 MIM: 610081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM9 PubMed Links

Gene Details

Gene
ADAM9
Gene Name
ADAM metallopeptidase domain 9
There are no transcripts associated with this gene.

Gene
HTRA4
Gene Name
HtrA serine peptidase 4
There are no transcripts associated with this gene.

Gene
TM2D2
Gene Name
TM2 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024380.1 912 Silent Mutation ACA,ACG T139T NP_001019551.1
NM_001024381.1 912 Silent Mutation ACA,ACG T139T NP_001019552.1
NM_031940.3 912 Silent Mutation ACA,ACG T139T NP_114146.3
NM_078473.2 912 Silent Mutation ACA,ACG T182T NP_510882.1
XM_005273657.3 912 Silent Mutation ACA,ACG T139T XP_005273714.1
XM_006716408.3 912 Silent Mutation ACA,ACG T139T XP_006716471.1
XM_011544667.2 912 Silent Mutation ACA,ACG T139T XP_011542969.1

View Full Product Details