Product Details

SNP ID
rs201203457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22691072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGGTTGTAGTCAGGAATCATGG[C/G]GAAGAGATTGCTGTCCAACGCCGGC
Phenotype
MIM: 602419
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EGR3 PubMed Links

Gene Details

Gene
EGR3
Gene Name
early growth response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199880.1 874 Missense Mutation CCC,GCC P151A NP_001186809.1
NM_001199881.1 874 Missense Mutation CCC,GCC P135A NP_001186810.1
NM_004430.2 874 Missense Mutation CCC,GCC P189A NP_004421.2
XM_005273425.3 874 Missense Mutation CCC,GCC P244A XP_005273482.1
XM_005273426.3 874 Missense Mutation CCC,GCC P151A XP_005273483.1
XM_011544429.2 874 Missense Mutation CCC,GCC P208A XP_011542731.1

View Full Product Details