Product Details

SNP ID
rs200505471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22248236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAAAAGAGAAGAGAGAGACAGA[C/T]GAAGAAACTAAACAGATCTTGCGTA
Phenotype
MIM: 614112 MIM: 187280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507071 PubMed Links

Gene Details

Gene
LOC100507071
Gene Name
uncharacterized LOC100507071
There are no transcripts associated with this gene.

Gene
MIR320A
Gene Name
microRNA 320a
There are no transcripts associated with this gene.

Gene
POLR3D
Gene Name
RNA polymerase III subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001722.2 527 Silent Mutation GAC,GAT D148D NP_001713.2

View Full Product Details