Product Details

SNP ID
rs199656487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:8319231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGACGAAGTGGCCGCAGTCCTG[A/C]TGGATGTTAAAGTGCACGGGCACGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SGK223 PubMed Links

Gene Details

Gene
SGK223
Gene Name
homolog of rat pragma of Rnd2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080826.2 3819 Missense Mutation CAG,CAT Q1048H NP_001074295.2
XM_005272369.4 3819 Missense Mutation CAG,CAT Q1048H XP_005272426.2
XM_005272370.4 3819 Missense Mutation CAG,CAT Q1048H XP_005272427.2
XM_011543813.2 3819 Intron XP_011542115.1

View Full Product Details