Product Details

SNP ID
rs199974181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:243006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGAGAACATCAGTCATCTGGT[C/G]TCTATTGGTGAGTCTCTTTATATTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF596 PubMed Links

Gene Details

Gene
ZNF596
Gene Name
zinc finger protein 596
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042415.2 420 Silent Mutation GTC,GTG V44V NP_001035880.1
NM_001042416.2 420 Silent Mutation GTC,GTG V44V NP_001035881.1
NM_001287254.1 420 Silent Mutation GTC,GTG V44V NP_001274183.1
NM_001287255.1 420 Silent Mutation GTC,GTG V44V NP_001274184.1
NM_001287256.1 420 Silent Mutation GTC,GTG V44V NP_001274185.1
NM_001287399.1 420 UTR 5 NP_001274328.1
NM_173539.3 420 Silent Mutation GTC,GTG V44V NP_775810.2
XM_011534744.1 420 Silent Mutation GTC,GTG V9V XP_011533046.1
XM_017013166.1 420 Silent Mutation GTC,GTG V44V XP_016868655.1

View Full Product Details