Product Details
- SNP ID
-
rs200611785
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143043193 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACGCACAGGGCCCAGGGGCTGCT[A/G]GCTGCCAGGACCACCGCATTGCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C8orf31
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11136300] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C8orf31
- Gene Name
- chromosome 8 open reading frame 31
- Gene
- LOC107984097
- Gene Name
- uncharacterized protein FLJ40521
There are no transcripts associated with this gene.
View Full Product Details