Product Details

SNP ID
rs200828547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143866668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCGTTTCCAGGTCCTGGAGCA[C/T]TTCCTCCGTGATTATGGCTGAGCTG
Phenotype
MIM: 607553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPPK1 PubMed Links

Gene Details

Gene
EPPK1
Gene Name
epiplakin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031308.3 6829 Missense Mutation ATG,GTG M2196V NP_112598.3
XM_017013889.1 6829 Missense Mutation ATG,GTG M2229V XP_016869378.1
XM_017013890.1 6829 Missense Mutation ATG,GTG M2196V XP_016869379.1
XM_017013891.1 6829 Missense Mutation ATG,GTG M1268V XP_016869380.1
XM_017013892.1 6829 Missense Mutation ATG,GTG M2229V XP_016869381.1

View Full Product Details