Product Details

SNP ID
rs200238621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42398788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTAGAAACCAAATATAAGGTCT[A/G]TAACTATGGACTTACCTTCACCCAG
Phenotype
MIM: 605417 MIM: 610029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKK4 PubMed Links

Gene Details

Gene
DKK4
Gene Name
dickkopf WNT signaling pathway inhibitor 4
There are no transcripts associated with this gene.

Gene
VDAC3
Gene Name
voltage dependent anion channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135694.2 407 Missense Mutation TAT,TGT Y66C NP_001129166.1
NM_005662.6 407 Missense Mutation TAT,TGT Y65C NP_005653.3
XM_006716394.1 407 Missense Mutation TAT,TGT Y33C XP_006716457.1

View Full Product Details